Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TFEB Recombinant Rabbit Monoclonal Antibody (BLR070G)
Rabbit Recombinant Monoclonal Antibody
$173.10 - $452.00
Specifications
Antigen | TFEB |
---|---|
Clone | BLR070G |
Concentration | 100 μg/mL |
Content And Storage | 4°C |
Applications | Immunoprecipitation, Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA544331
|
Invitrogen™
MA544331 |
100 μL |
Each for $452.00
|
|
|||||
PIMA544332
|
Invitrogen™
MA544332 |
20 μL |
Each for $173.10
|
|
|||||
Description
TFEB Recombinant Monoclonal Antibody for Western Blot, IP
The Transcription factor EB (TFEB) is a member in the basic helix-loop-helix leucine zipper superfamily of transcription factors that is translocated in a subset of renal tumors. Recent studies have shown that lysosomal biogenesis is regulated by TFEB, which is in turn regulated by the mammalian target of rapamycin (mTOR) complex 1. Other evidence suggests that TFEB coordinates the major steps of the autophagic pathway by driving the expression of autophagy and lysosomal genes.Specifications
TFEB | |
100 μg/mL | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
BBS with 0.1% BSA and 0.09% sodium azide; pH 7.8-8.6 | |
P19484, Q9R210 | |
21425, 7942 | |
Peptide representing a region between residue 426 and the C-terminus (residue 476) of human Transcription factor EB. | |
Antibody |
BLR070G | |
4°C | |
Recombinant Monoclonal | |
Liquid | |
IgG | |
Human, Mouse | |
TFEB | |
Alpha TCFEB; ALPHATFEB; BHLHE35; Class E basic helix-loop-helix protein 35; RP4-696P19.3; T-cell transcription factor EB; Tcfeb; Tfeb; Transcription factor EB | |
TFEB | |
Primary | |
Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title