Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFF1/pS2 Antibody (TFF1/2969R), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB382368 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB382368 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB382368 Supplier Novus Biologicals Supplier No. NBP308252AF532
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TFF1/pS2 Monoclonal antibody specifically detects TFF1/pS2 in Human, Cynomolgus Monkey samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen TFF1/pS2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone TFF1/2969R
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias BCEIbreast cancer, estrogen-inducible sequence expressed in, Breast cancer estrogen-inducible protein, breast cancer estrogen-inducible sequence, D21S21, gastrointestinal trefoil protein pS2, hP1.A, HPS2, pNR-2, Polypeptide P1.A, Protein pS2, pS2, trefoil factor 1, trefoil factor, BCE1, human pS2 induced by estrogen from human breast cancercell line M10HP1.A
Host Species Rabbit
Immunogen Synthetic peptide of 28 amino acid residues corresponding to CFDDTVRGVPWCFYPNTIDVPPEEECEF (aa57-84) from the C-terminus of human TFF1/pS2 (Uniprot: P04155)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 7031
Target Species Human, Cynomolgus Monkey
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.