Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFF1/pS2 Mouse, Clone: GE2 (same as R47/94), Novus Biologicals™
SDP

Catalog No. NBP234293A Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
0.2 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP234293A 0.1 mg
NBP234293F 0.02 mg
NBP234293B 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP234293A Supplier Novus Biologicals Supplier No. NBP2342930.1MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TFF1/pS2 Monoclonal specifically detects TFF1/pS2 in Cynomolgus Monkey, Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen TFF1/pS2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone GE2 (same as R47/94)
Conjugate Unconjugated
Dilution Western Blot 0.5-1.0ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5-1ug/ml, SDS-Page
Formulation PBS with 0.05% BSA. with 0.05% Sodium Azide
Gene Accession No. P04155
Gene Alias BCEIbreast cancer, estrogen-inducible sequence expressed in, Breast cancer estrogen-inducible protein, breast cancer estrogen-inducible sequence, D21S21, gastrointestinal trefoil protein pS2, hP1.A, HPS2, pNR-2, Polypeptide P1.A, Protein pS2, pS2, trefoil factor 1, trefoil factor, BCE1, human pS2 induced by estrogen from human breast cancercell line M10HP1.A
Gene Symbols TFF1
Host Species Mouse
Immunogen Synthetic peptide of 28 amino acid residues corresponding to CFDDTVRGVPWCFYPNTIDVPPEEECEF (aa57-84) from the C-terminus of human pS2.
Molecular Weight of Antigen 6.5 kDa
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 7031
Test Specificity It recognizes a polypeptide of 6.5kDa, identified as pS2 estrogen-regulated protein. Its epitope is localized between aa57-84 of human pS2 protein. pS2 is a trefoil peptide. Trefoil peptides are protease resistant molecules secreted throughout the gut that play a role in mucosal healing. These peptides contain three intra-chain disulfide bonds, forming the trefoil motif, or P-domain. pS2 is known to form dimers and this dimerization is thought to play a role in its protective and healing properties. About 60% of breast carcinomas are positive for pS2. Staining is cytoplasmic, often with localization to the Golgi apparatus. pS2 is shown to be localized in normal stomach mucosa, gastric fluid, goblet cells in the colon and small intestine, and in ulcerations of the gastrointestinal tract. Several studies have shown that pS2 is primarily expressed in estrogen receptor-positive breast tumors and it may define a subset of estrogen-dependent tumors that displays an increased likelihood of response to endocrine therapy.
Target Species Cynomolgus Monkey, Human
Content And Storage Store at 4C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.