Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFF3 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP176514 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP176514 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP176514 Supplier Novus Biologicals Supplier No. NBP176514
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TFF3 Polyclonal specifically detects TFF3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence, Immunocytochemistry.

Specifications

Antigen TFF3
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 2 ug/ml, Immunocytochemistry/Immunofluorescence 2-20 ug/mL, Immunohistochemistry-Paraffin 2 ug/ml
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. NP_003217
Gene Alias HITF, human intestinal trefoil factor10Intestinal trefoil factor, trefoil factor 3 (intestinal)
Gene Symbols TFF3
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of human TFF3. The immunogen is located within the last 50 amino acids of TFF3. Amino Acid Squence: ECNNRGCCFDSRIPGV
Molecular Weight of Antigen 11 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 7033
Test Specificity TFF3 antibody is predicted to not cross-react with other TFF family members.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.