Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TFF3 Mouse anti-Human, Unconjugated, Clone: TFF-286, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABS2053100UG
Description
TFF3 Monoclonal antibody specifically detects TFF3 in Human samples. It is validated for ELISA, Western Blotting.
Specifications
TFF3 | |
Monoclonal | |
Unconjugated | |
Trefoil factor 3;Intestinal trefoil factor;hITF;Polypeptide P1.B;hP1.B | |
Mouse | |
∼26 kDa observed for His-and thioredoxin tagged recombinant human TFF3; 10.18 kDa calculated. Uncharacterized bands may be observed in some lysate(s). | |
100 μg | |
Primary | |
Human | |
Purified |
ELISA, Western Blot | |
TFF-286 | |
NP_003217 | |
TFF3, ITF, TFI | |
Recombinant human dimeric TFF3. | |
Protein G Purified | |
Signaling | |
Clone TFF-286 is a mouse monoclonal antibody that detects human Trefoil factor 3 (TFF3). | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction