Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TFF3 Mouse anti-Human, Unconjugated, Clone: TFF-286, MilliporeSigma™
Mouse Monoclonal Antibody
$182.31 - $457.74
Specifications
Antigen | TFF3 |
---|---|
Clone | TFF-286 |
Applications | ELISA, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABS20532MI
|
MilliporeSigma
MABS205325UG |
25 μg |
Each for $182.31
|
|
|||||
MABS20531MI
|
MilliporeSigma
MABS2053100UG |
100 μg |
Each for $457.74
|
|
|||||
Description
TFF3 Monoclonal antibody specifically detects TFF3 in Human samples. It is validated for ELISA, Western Blotting.Specifications
TFF3 | |
ELISA, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Trefoil factor 3;Intestinal trefoil factor;hITF;Polypeptide P1.B;hP1.B | |
Recombinant human dimeric TFF3. | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
∼26 kDa observed for His-and thioredoxin tagged recombinant human TFF3; 10.18 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
TFF-286 | |
Monoclonal | |
Purified | |
Signaling | |
NP_003217 | |
TFF3, ITF, TFI | |
IgG1 κ | |
Protein G Purified | |
Clone TFF-286 is a mouse monoclonal antibody that detects human Trefoil factor 3 (TFF3). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title