Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFG Antibody (TFG-03), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB002557 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002557 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002557 Supplier Novus Biologicals Supplier No. NBP262212R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TFG Monoclonal specifically detects TFG in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen TFG
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone TFG-03
Conjugate DyLight 550
Dilution Western Blot, Immunocytochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FLJ36137, protein TFG, TF6, TRK-fused gene, TRK-fused gene protein, TRKT3, TRKT3 oncogene
Gene Symbols TFG
Host Species Mouse
Immunogen Human recombinant protein Trx-his-NTFG
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10342
Test Specificity The antibody MEM-57 reacts with gamma-epsilon and delta-epsilon dimers of human CD3 complex, a part of a bigger multisubunit T cell receptor complex (CD3/TCR) expressed on peripheral blood T lymphocytes and mature thymocytes.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.