Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFG Antibody (TFG-03), PerCP, Novus Biologicals™
SDP

Catalog No. NB343078 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343078 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343078 Supplier Novus Biologicals Supplier No. NBP262212PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TFG Monoclonal antibody specifically detects TFG in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Immunocytochemistry

Specifications

Antigen TFG
Applications Western Blot, Flow Cytometry, Immunoprecipitation, Immunocytochemistry
Classification Monoclonal
Clone TFG-03
Conjugate PerCP
Dilution Western Blot, Flow Cytometry, Immunoprecipitation, Immunocytochemistry
Formulation PBS
Gene Alias FLJ36137, protein TFG, TF6, TRK-fused gene, TRK-fused gene protein, TRKT3, TRKT3 oncogene
Host Species Mouse
Immunogen Human recombinant protein Trx-his-NTFG
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10342
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.