Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFIIB Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB291312 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291312 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291312 Supplier Novus Biologicals Supplier No. NBP149981F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TFIIB Polyclonal antibody specifically detects TFIIB in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)

Specifications

Antigen TFIIB
Applications Immunoprecipitation
Classification Polyclonal
Conjugate FITC
Dilution Immunoprecipitation, Western Blot (Negative)
Formulation PBS
Gene Alias general transcription factor IIB, General transcription factor TFIIB, S300-II, TFIIBTF2BRNA polymerase II transcription factor IIB, transcription initiation factor IIB
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 100 and 150 of human General Transcription Factor IIB using the numbering given in entry NP_001505.1 (GeneID 2959).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 2959
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.