Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFIIB Antibody (OTI1H10), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP274492A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP274492A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP274492A32 Supplier Novus Biologicals Supplier No. NBP274492AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TFIIB Monoclonal specifically detects TFIIB in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen TFIIB
Applications Western Blot
Classification Monoclonal
Clone OTI1H10
Conjugate Alexa Fluor 532
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias general transcription factor IIB, General transcription factor TFIIB, S300-II, TFIIBTF2BRNA polymerase II transcription factor IIB, transcription initiation factor IIB
Gene Symbols GTF2B
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 43-316 of human TFIIB (NP_001505) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 2959.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.