Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFIIB Mouse anti-Human, Clone: PCRP-GTF2B-1D1, Novus Biologicals™
SDP

Catalog No. NB177825 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB177825 20 μg
NB177824 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB177825 Supplier Novus Biologicals Supplier No. NBP31393420UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TFIIB Monoclonal antibody specifically detects TFIIB in Human samples. It is validated for Flow Cytometry, ELISA, Immunoprecipitation, Protein Array

Specifications

Antigen TFIIB
Applications Flow Cytometry, ELISA, Immunoprecipitation, Peptide Array
Classification Monoclonal
Clone PCRP-GTF2B-1D1
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry 1-2 ug/million cells, ELISA, Immunoprecipitation 1-2 ug/100-500 ug protein, Protein Array
Formulation 10 mM PBS with 0.05% BSA
Gene Accession No. Q00403
Gene Alias general transcription factor IIB, General transcription factor TFIIB, S300-II, TFIIBTF2BRNA polymerase II transcription factor IIB, transcription initiation factor IIB
Host Species Mouse
Immunogen Recombinant full-length human TFIIB protein (Uniprot: Q00403)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline DNA Replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 2959
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.