Learn More
Description
Specifications
Specifications
| Antigen | TFIIE-alpha |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:2000-1:10000, Immunohistochemistry, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:100 - 1:500 |
| Formulation | TBS and 0.1% BSA with 0.09% Sodium Azide |
| Gene Alias | FE, General transcription factor IIE 56 kDa subunit, general transcription factor IIE subunit 1, general transcription factor IIE, polypeptide 1 (alpha subunit, 56kD), general transcription factor IIE, polypeptide 1, alpha 56kDa, TF2E1, TFIIE-A, TFIIE-alpha, Transcription initiation factor IIE subunit alpha |
| Gene Symbols | GTF2E1 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
