Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFIIE beta Antibody, Novus Biologicals™
SDP

Catalog No. NBP187931 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP187931 0.1 mL
NB418426 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP187931 Supplier Novus Biologicals Supplier No. NBP187931
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

TFIIE beta Polyclonal specifically detects TFIIE beta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen TFIIE beta
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias FE, General transcription factor IIE subunit 2, general transcription factor IIE, polypeptide 2 (beta subunit, 34kD), general transcription factor IIE, polypeptide 2, beta 34kDa, TF2E2TFIIE-beta, TFIIE-B, transcription initiation factor IIE subunit beta
Gene Symbols GTF2E2
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:PSLLRERELFKKRALSTPVVEKRSASSESSSSSSKKKKTKVEHGGSSGSKQNSDHSNGSFNLKALSGSSGYKFGVL
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 2961
Test Specificity Specificity of human TFIIE beta antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.