Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFIIIC Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB291618 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291618 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291618 Supplier Novus Biologicals Supplier No. NB10060657PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TFIIIC Polyclonal specifically detects TFIIIC in Human, Mouse samples. It is validated for ChIP Assay, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation (Negative), Western Blot.

Specifications

Antigen TFIIIC
Applications ChIP Assay, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation (Negative), Western Blot
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoprecipitation (Negative), Chromatin Immunoprecipitation (ChIP)
Formulation PBS
Gene Alias general transcription factor 3C polypeptide 1, general transcription factor IIIC, polypeptide 1 (alpha subunit), general transcription factor IIIC, polypeptide 1 (alpha subunit, 220kD ), general transcription factor IIIC, polypeptide 1, alpha 220kDa, TF3C-alpha, TFIIIC, TFIIIC 220 kDa subunit, TFIIIC box B-binding subunit, TFIIIC220DKFZp686A111, TFIIICalpha, Transcription factor IIIC 220 kDa subunit, Transcription factor IIIC subunit alpha
Host Species Rabbit
Immunogen Maps to a region between residue 2059 and 2109 of human general transcription factor IIIC, polypeptide 1 (general transcription factor IIIC, 220kDa subunit) using the numbering given in entry NP_001511.2 (GeneID 2
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2975
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.