Learn More
TFIIS Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | TFIIS |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-2482
![]() |
Bethyl Laboratories, Inc
IHC00656T |
10 μL |
Each for $151.31
|
|
|||||
50-157-2481
![]() |
Bethyl Laboratories, Inc
IHC00656 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Monkey, Gorilla, Chimpanzee, Northern white-cheeked gibbon, Crab-eating macaque and Wh
TCEA1 is necessary for efficient RNA polymerase II transcription elongation past template-encoded arresting sites. TCEA1 has a role in suppression of transient pausing, which is the most important contribution of TCEA1 to elongation from a stall position. TCEA1 fuse to PLAG1 protein which is overexpressed in epithelial, myoepithelial, and mesenchymal-like tumor cells in tumors.Specifications
TFIIS | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Transcription elongation factor A (SII), 1, SII, TCEA, TF2S, GTF2S, TFIIS | |
TCEA1 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
P23193 | |
6917 | |
Between 1 and 50 | |
Primary | |
4° C | |
TCEA1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.