Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFPI-2 Antibody (OTI1D4), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP274494A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP274494A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP274494A32 Supplier Novus Biologicals Supplier No. NBP274494AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TFPI-2 Monoclonal specifically detects TFPI-2 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen TFPI-2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1D4
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Placental protein 5, PP5FLJ21164, TFPI-2REF1, tissue factor pathway inhibitor 2
Gene Symbols TFPI2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 23-235 of human TFPI-2 (NP_006519) produced in E.coli.
Quantity 0.1 mL
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 7980
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.