Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFPI Antibody (107), Alexa Fluor™ 532
SDP

Catalog No. NB073526 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB073526 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB073526 Supplier Novus Biologicals Supplier No. NBP290440AF532

Rabbit Monoclonal Antibody

TFPI Monoclonal specifically detects TFPI in Mouse samples. It is validated for ELISA.

Specifications

Antigen TFPI
Applications ELISA
Classification Monoclonal
Clone 107
Conjugate Alexa Fluor 532
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias convertin, EPITFPI1LACITFI, Extrinsic pathway inhibitor, Lipoprotein-associated coagulation inhibitor, tissue factor pathway inhibitor, tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor)
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TFPI (NP_035706.1; Met 1-Lys 289).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 7035
Test Specificity No cross-reactivity in ELISA with: Human TFPI
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.