Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFPI Antibody (6), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB108388 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB108388 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB108388 Supplier Novus Biologicals Supplier No. NBP289569AF750
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TFPI Monoclonal specifically detects TFPI in Human samples. It is validated for Western Blot.

Specifications

Antigen TFPI
Applications Western Blot
Classification Monoclonal
Clone 6
Conjugate Alexa Fluor 750
Formulation 50 mM Sodium Borate
Gene Alias convertin, EPITFPI1LACITFI, Extrinsic pathway inhibitor, Lipoprotein-associated coagulation inhibitor, tissue factor pathway inhibitor, tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor)
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human TFPI (NP_006278.1; Met 1-Lys 282).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 7035
Test Specificity No cross-reactivity in ELISA with: Mouse TFPI/ LACI/ EPI Human TFPI2/ PP5 Human Vitronectin/ VTN
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.