Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFPI Antibody (OTI5H5), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP274493V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274493V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274493V Supplier Novus Biologicals Supplier No. NBP274493V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TFPI Monoclonal specifically detects TFPI in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen TFPI
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI5H5
Conjugate DyLight 405
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias convertin, EPITFPI1LACITFI, Extrinsic pathway inhibitor, Lipoprotein-associated coagulation inhibitor, tissue factor pathway inhibitor, tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor)
Gene Symbols TFPI
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 29-304 of human TFPI (NP_006278) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 7035.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.