Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TfR (Transferrin R) Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB003835 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB003835 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB003835 Supplier Novus Biologicals Supplier No. NB10092243AF405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TfR (Transferrin R) Polyclonal specifically detects TfR (Transferrin R) in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Proximity Ligation Assay, Western Blot.

Specifications

Antigen TfR (Transferrin R)
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Proximity Ligation Assay, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD71 antigen, CD71TRFR, p90, T9, TfR, TFR1, TR, transferrin receptor (p90, CD71), transferrin receptor protein 1, Trfr
Gene Symbols TFRC
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human Transferrin R (between amino acids 20-75) [UniProt P02786]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.