Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TfR (Transferrin R) Antibody (OX26), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB200585X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB200585X 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB200585X Supplier Novus Biologicals Supplier No. NB200585AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TfR (Transferrin R) Monoclonal specifically detects TfR (Transferrin R) in Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular).

Specifications

Antigen TfR (Transferrin R)
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone OX26
Conjugate Alexa Fluor 488
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular)
Gene Alias CD71 antigen, CD71TRFR, p90, T9, TfR, TFR1, TR, transferrin receptor (p90, CD71), transferrin receptor protein 1, Trfr
Gene Symbols TFRC
Host Species Mouse
Immunogen PHA activated rat lymphocytes.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7037
Test Specificity Labels dividing cells of various types but also binds to a number of non-dividing normal tissues and endothelium of brain capillaries.
Target Species Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.