Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TfR (Transferrin R) Rabbit anti-Human, Clone: 1, Novus Biologicals™
SDP

Catalog No. NB020966 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB020966 50 μg
NB020965 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB020966 Supplier Novus Biologicals Supplier No. NBP28974250UL

Rabbit Monoclonal Antibody

TfR (Transferrin R) Monoclonal specifically detects TfR (Transferrin R) in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Knockout Validated.

Specifications

Antigen TfR (Transferrin R)
Applications Western Blot, ELISA, Immunoprecipitation, KnockDown
Classification Monoclonal
Clone 1
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 0.5-2 ul/mg of lysate, Knockout Validated
Formulation 0.2 um filtered solution in PBS
Gene Alias CD71 antigen, CD71TRFR, p90, T9, TfR, TFR1, TR, transferrin receptor (p90, CD71), transferrin receptor protein 1, Trfr
Host Species Rabbit
Immunogen Recombinant Human TfR (Transferrin R) protein
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7037
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.