Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ TG 003

Click to view available options
Quantity:
10 mg
50 mg
Description
Potent, ATP-competitive inhibitor of Clk-family kinases (IC50 values are 15, 20 and 200 nM for mClk4, 1 and 2 respectively and >10 μM for mClk3). Also inhibits DYRK1A/B (IC50 values are 24 and 34 nM respectively). Suppresses serine/arginine-rich protein phosphorylation; affects the regulation of alternative splicing by phosphorylation of SR protein both in vitro and in vivo.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 249.33 |
| Purity | >98% |
| Molecular Weight (g/mol) | 249.328 |
| Chemical Name or Material | TG 003 |
Safety and Handling
| Recommended Storage | Store at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction