Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF-alpha Antibody (SPM357), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB382183 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB382183 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB382183 Supplier Novus Biologicals Supplier No. NBP308270AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TGF-alpha Monoclonal antibody specifically detects TGF-alpha in Human, Mouse, Rat, Bovine samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen TGF-alpha
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SPM357
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias protransforming growth factor alpha, TFGA, transforming growth factor, alpha, transforming growth factor-alpha
Host Species Mouse
Immunogen A 17-amino acid synthetic peptide from the carboxyl-terminus of rat TGF-alpha (Uniprot: P01135)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Cell Cycle and Replication, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7039
Target Species Human, Mouse, Rat, Bovine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.