Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF-alpha Antibody (tAb2), HRP, Novus Biologicals™
SDP

Catalog No. NB015634 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB015634 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB015634 Supplier Novus Biologicals Supplier No. NBP281082H
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TGF-alpha Monoclonal specifically detects TGF-alpha in Human samples. It is validated for ELISA, Dot Blot.

Specifications

Antigen TGF-alpha
Applications ELISA, Dot Blot
Classification Monoclonal
Clone tAb2
Conjugate HRP
Dilution ELISA, Dot Blot
Formulation PBS
Gene Alias protransforming growth factor alpha, TFGA, transforming growth factor, alpha, transforming growth factor-alpha
Gene Symbols TGFA
Host Species Rabbit
Immunogen Biologically active hemocyanin-coupled human TGF-alpha.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cell Cycle and Replication, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7039
Test Specificity tAb2 binds the N-terminal sequence of TGF-alpha, VVSHFND (with residues 4 to 7 being the minimal binding epitope).
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.