Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF-alpha Antibody (tAb2), PerCP, Novus Biologicals™
SDP

Catalog No. NB342849 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342849 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342849 Supplier Novus Biologicals Supplier No. NBP281082PCP
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TGF-alpha Monoclonal antibody specifically detects TGF-alpha in Human samples. It is validated for Flow Cytometry, ELISA, Dot Blot

Specifications

Antigen TGF-alpha
Applications Flow Cytometry, ELISA, Dot Blot
Classification Monoclonal
Clone tAb2
Conjugate PerCP
Dilution Flow Cytometry, ELISA, Dot Blot
Formulation PBS
Gene Alias protransforming growth factor alpha, TFGA, transforming growth factor, alpha, transforming growth factor-alpha
Host Species Rabbit
Immunogen Biologically active hemocyanin-coupled human TGF-alpha.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cell Cycle and Replication, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7039
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.