Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF-alpha Mouse, Clone: P/T1, Novus Biologicals™
SDP

Catalog No. NBP234299A Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
0.2 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP234299A 0.1 mg
NBP234299F 0.02 mg
NBP234299B 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP234299A Supplier Novus Biologicals Supplier No. NBP2342990.1MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TGF-alpha Monoclonal specifically detects TGF-alpha in Human, Rabbit, Zebrafish samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen TGF-alpha
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone P/T1
Conjugate Unconjugated
Dilution Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunoprecipitation 1-2ug/500ug protein, Immunohistochemistry-Paraffin 1-2ug/ml, Immunohistochemistry-Frozen 1-2ug/ml
Formulation PBS with 0.05% BSA. with 0.05% Sodium Azide
Gene Accession No. P01135
Gene Alias protransforming growth factor alpha, TFGA, transforming growth factor, alpha, transforming growth factor-alpha
Gene Symbols TGFA
Host Species Mouse
Immunogen A 10-amino acid synthetic peptide (aa 34-43; PPVAAAVVSH) from human TGF alpha.
Molecular Weight of Antigen 6 kDa
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Angiogenesis, Cell Cycle and Replication, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7039
Test Specificity This antibody reacts with the TGF alpha and shows no cross-reaction with EGF and the neuropeptide synenkephalin. The staining with this MAb is completely blocked by the peptide used for raising this antibody.
Target Species Human, Rabbit, Zebrafish
Content And Storage Store at 4C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.