Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TGF-alpha Mouse, Clone: P/T1, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $524.00
Specifications
Antigen | TGF-alpha |
---|---|
Clone | P/T1 |
Dilution | Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunoprecipitation 1-2ug/500ug protein, Immunohistochemistry-Paraffin 1-2ug/ml, Immunohistochemistry-Frozen 1-2ug/ml |
Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP234299F
![]() |
Novus Biologicals
NBP2342990.02MG |
0.02 mg |
Each for $268.00
|
|
|||||
NBP234299A
![]() |
Novus Biologicals
NBP2342990.1MG |
0.1 mg |
Each for $524.00
|
|
|||||
NBP234299B
![]() |
Novus Biologicals
NBP2342990.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
TGF-alpha Monoclonal specifically detects TGF-alpha in Human, Rabbit, Zebrafish samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TGF-alpha | |
Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunoprecipitation 1-2ug/500ug protein, Immunohistochemistry-Paraffin 1-2ug/ml, Immunohistochemistry-Frozen 1-2ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human, Rabbit, Zebrafish | |
P01135 | |
7039 | |
A 10-amino acid synthetic peptide (aa 34-43; PPVAAAVVSH) from human TGF alpha. | |
Primary | |
Store at 4C. | |
6 kDa |
P/T1 | |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Angiogenesis, Cell Cycle and Replication, Signal Transduction | |
PBS with 0.05% BSA. with 0.05% Sodium Azide | |
protransforming growth factor alpha, TFGA, transforming growth factor, alpha, transforming growth factor-alpha | |
TGFA | |
IgG1 κ | |
Protein A purified | |
This antibody reacts with the TGF alpha and shows no cross-reaction with EGF and the neuropeptide synenkephalin. The staining with this MAb is completely blocked by the peptide used for raising this antibody. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title