Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF-alpha Rabbit anti-Human, Clone: 12, Novus Biologicals™
SDP

Catalog No. NB021188 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB021188 100 μg
NB021189 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB021188 Supplier Novus Biologicals Supplier No. NBP289861100UL

Rabbit Monoclonal Antibody

TGF-alpha Monoclonal specifically detects TGF-alpha in Human samples. It is validated for ELISA.

Specifications

Antigen TGF-alpha
Applications ELISA
Classification Monoclonal
Clone 12
Conjugate Unconjugated
Dilution ELISA 1:5000-1:10000
Formulation 0.2 um filtered solution in PBS
Gene Alias protransforming growth factor alpha, TFGA, transforming growth factor, alpha, transforming growth factor-alpha
Host Species Rabbit
Immunogen Recombinant Human TGF-alpha Protein
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Cell Cycle and Replication, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7039
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.