Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF beta 1/2/3 Antibody (1D11), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB367394 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB367394 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB367394 Supplier Novus Biologicals Supplier No. FAB1835S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TGF beta 1/2/3 Monoclonal specifically detects TGF beta 1/2/3 in Multi-species samples. It is validated for Binding Inhibition, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), ELISA Standard (Matched Pair), Neutralization, Western Blot.

Specifications

Antigen TGF beta 1/2/3
Applications Binding Inhibition Assay, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), ELISA Standard (Matched Pair), Neutralization, Western Blot
Classification Monoclonal
Clone 1D11
Conjugate Alexa Fluor 750
Dilution Western Blot, Binding Inhibition, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair)
Formulation 50mM Sodium Borate
Gene Alias CED, DPD1, LAP, TGFB, TGFB1, TGFbeta, transforming growth factor, beta 1
Host Species Mouse
Immunogen Bovine bone-derived TGF-beta 1 and TGF-beta 2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7040
Target Species Multi-species
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.