Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF-beta 1 Antibody (1018746) [DyLight 405], Novus Biologicals™
SDP

Catalog No. NB401486 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB401486 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB401486 Supplier Novus Biologicals Supplier No. FAB10502E
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TGF-beta 1 Monoclonal antibody specifically detects TGF-beta 1 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF

Specifications

Antigen TGF-beta 1
Applications Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 1018746
Conjugate DyLight 405
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CEDLAP, DPD1, latency-associated peptide, TGF beta, TGFB, TGFbeta, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1
Host Species Mouse
Immunogen Chinese Hamster Ovary cell line, CHO-derived human TGF-beta 1 protein, Ala279-Ser390, Accession # P01137
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Apoptosis, Asthma, Cancer, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7040
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.