Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF-beta 1 Antibody (MM0446-6G14), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NBP211766Y Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211766Y 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP211766Y Supplier Novus Biologicals Supplier No. NBP211766AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TGF-beta 1 Monoclonal specifically detects TGF-beta 1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen TGF-beta 1
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone MM0446-6G14
Conjugate Alexa Fluor 647
Dilution Western Blot, Flow Cytometry
Gene Accession No. P01137
Gene Alias CEDLAP, DPD1, latency-associated peptide, TGFB, TGFbeta, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1
Gene Symbols TGFB1
Host Species Mouse
Immunogen Human recombinant TGF-beta 1
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Apoptosis, Asthma, Cancer, Immunology, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 7040
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.