Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF-beta 1 Antibody (MM0446-6G14), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NBP21766549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP21766549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP21766549 Supplier Novus Biologicals Supplier No. NBP211766JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TGF-beta 1 Monoclonal specifically detects TGF-beta 1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen TGF-beta1
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone MM0446-6G14
Conjugate Janelia Fluor 549
Dilution Western Blot, Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CEDLAP, DPD1, latency-associated peptide, TGFB, TGFbeta, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1
Gene Symbols TGFB1
Host Species Mouse
Immunogen Human recombinant TGF-beta 1
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Angiogenesis, Apoptosis, Asthma, Cancer, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7040
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.