Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse TGF-beta 1 ELISA Kit (Colorimetric)
SDP

Catalog No. NBP192671 Shop All R&D Systems Products
Click to view available options
Quantity:
96 Tests

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The Mouse TGF-beta 1 ELISA Kit (Colorimetric) measures mouse TGF-beta 1 in cell culture supernatant, serum, plasma (edta, citrate).

Specifications

Assay Range 31.3 - 2,000pg/mL
Assay Sensitivity 9.0pg/mL
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Plasma (EDTA, Citrate), Serum
For Use With (Application) ELISA
Gene ID (Entrez) 7040
Gene Symbol TGFB1
Quantity 96 Tests
Research Discipline Angiogenesis, Apoptosis, Asthma, Cancer, Immunology
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym CEDLAP, DPD1, latency-associated peptide, TGFB, TGFbeta, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1
Target TGF-beta 1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.