Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TGF beta-3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578197
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578197 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578197 Supplier Invitrogen™ Supplier No. PA578197
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: human-TGFB3 transfected 293T Predicted Reactivity: Rat (97%), Pig (91%), Chicken (87%), Rhesus Monkey (100%), Bovine (87%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Transforming growth factor beta 3 (TGF beta 3) is the third member of the transforming growth factor family of cytokines, which also includes TGF beta 1 and beta 2. These cytokines are secreted in precursor form consisting of a bioactive C-terminal domain attached to an N-terminal domain known as latency associated protein (LAP). Cleavage of LAP results in the mature protein, which functions as a disulfide-linked homodimer. As with all members of the family, TGF beta 3 is highly conserved across species, with mouse and human TGF beta 3 demonstrating 100% sequence homology and cross-species activity.The members of this family can be expressed by most cell types and exert pleiotropic effects, which include the suppression of B- and T-cell effector activity, mediation of tissue healing, and suppression of tumor proliferation. The promotion of CD4+CD25+ T-cell expansion is a newly discovered function of the TGF beta cytokines, and indicates an important role in the protection against autoimmunity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TGF beta-3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene TGFB3
Gene Accession No. P10600, P17125
Gene Alias ARVD; ARVD1; FLJ16571; H-TGF-b-3; LAP; Latency-associated peptide; LDS5; prepro-transforming growth factor beta-3; RNHF; TGF beta 3; Tgfb3; TGF-B3; Tgfb-3; TGF-beta3; TGF-beta-3; Transforming growth factor; transforming growth factor beta 3; transforming growth factor beta precursor; transforming growth factor beta-3; Transforming growth factor beta-3 proprotein; transforming growth factor, beta 3; transforming growth factor-beta3
Gene Symbols TGFB3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human TGF beta 3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21809, 7043
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.