Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF-beta Antibody (TGFB/510), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB391060 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB391060 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB391060 Supplier Novus Biologicals Supplier No. NBP308739FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TGF-beta Monoclonal antibody specifically detects TGF-beta in Human, Mouse, Rat, Bovine, Canine, Hamster, Monkey samples. It is validated for ELISA, Functional Assay

Specifications

Antigen TGF-beta
Applications ELISA, Functional Assay
Classification Monoclonal
Clone TGFB/510
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias CED, DPD1, LAP, TGFB, TGFB1, TGFbeta, transforming growth factor beta 1
Host Species Mouse
Immunogen Full length recombinant human TGF-beta protein
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cytokine Research, Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 7040
Target Species Human, Mouse, Rat, Bovine, Canine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.