Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF-beta RI/ALK-5 Antibody (RM0016-3A11), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB211921647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB211921647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB211921647 Supplier Novus Biologicals Supplier No. NBP211921AF647
Add to Cart
Add to Cart

Rat Monoclonal Antibody

TGF-beta RI/ALK-5 Monoclonal specifically detects TGF-beta RI/ALK-5 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen TGF-beta RI/ALK-5
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RM0016-3A11
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AAT5, activin A receptor type II-like kinase, 53kD, activin A receptor type II-like kinase, 53kDa, Activin receptor-like kinase 5, ACVRLK4, ALK-5ALK5, EC 2.7.11, EC 2.7.11.30, LDS1A, LDS2A, Serine/threonine-protein kinase receptor R4, SKR4, tbetaR-I, TGFB1R1, TGF-beta receptor type I, TGF-beta receptor type-1, TGF-beta type I receptor, TGFR-1, transforming growth factor beta receptor I, transforming growth factor, beta receptor 1, transforming growth factor, beta receptor I (activin A receptor type II-likekinase, 53kD), Transforming growth factor-beta receptor type I
Gene Symbols TGFBR1
Host Species Rat
Immunogen Mouse TGF-beta RI/ALK-5 (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Angiogenesis, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 7046
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.