Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF-beta RII Antibody (RM0132-6G11), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP122508V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP122508V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP122508V Supplier Novus Biologicals Supplier No. NBP122508V
Add to Cart
Add to Cart

Rat Monoclonal Antibody

TGF-beta RII Monoclonal specifically detects TGF-beta RII in Mouse samples. It is validated for Western Blot.

Specifications

Antigen TGF-beta RII
Applications Western Blot
Classification Monoclonal
Clone RM0132-6G11
Conjugate DyLight 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AAT3, EC 2.7.11, EC 2.7.11.30, FAA3, HNPCC6, LDS1B, LDS2B, MFS2, RIIC, TAAD2, tbetaR-II, TGF-beta receptor type II, TGF-beta receptor type IIB, TGF-beta receptor type-2, TGF-beta type II receptor, TGFbeta-RII, TGFR-2, transforming growth factor beta receptor type IIC, transforming growth factor, beta receptor II (70/80kDa), transforming growth factor, beta receptor II (70/80kDa) isoform 1, transforming growth factor, beta receptor II (70/80kDa) isoform 2, transforming growth factor, beta receptor II (70-80kD), Transforming growth factor-beta receptor type II
Gene Symbols TGFBR2
Host Species Rat
Immunogen Mouse recombinant TGF-beta RII (extracellular domain)
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Angiogenesis, Cell Biology, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 7048
Test Specificity This antibody does not cross-react with mouse TGF-beta RI.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.