Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF-beta RIII Antibody (030), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB075842 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB075842 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB075842 Supplier Novus Biologicals Supplier No. NBP290633AF350
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TGF-beta RIII Monoclonal specifically detects TGF-beta RIII in Mouse samples. It is validated for ELISA.

Specifications

Antigen TGF-beta RIII
Applications ELISA
Classification Monoclonal
Clone 30
Conjugate Alexa Fluor 350
Formulation 50 mM Sodium Borate
Gene Alias betaglycan, betaglycan proteoglycan, BGCAN, TGF-beta receptor type 3, TGF-beta receptor type III, TGFR-3, Transforming growth factor beta receptor III, transforming growth factor beta receptor type 3, transforming growth factor, beta receptor III, transforming growth factor, beta receptor III (betaglycan, 300kDa)
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TGF-beta RIII (NP_072075.2; Met1-Arg399).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis
Primary or Secondary Primary
Gene ID (Entrez) 7049
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.