Learn More
Description
Specifications
Specifications
| Antigen | TGF-betaRIII |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000, Immunohistochemistry 5-10 ug/ml, Immunoprecipitation 5 ug/mg protein lysate, Immunohistochemistry-Paraffin 1-5 ug/ml |
| Formulation | PBS with 0.08% Sodium Azide |
| Gene Accession No. | P16047 |
| Gene Alias | betaglycan, betaglycan proteoglycan, BGCAN, TGF-beta receptor type 3, TGF-beta receptor type III, TGFR-3, Transforming growth factor beta receptor III, transforming growth factor beta receptor type 3, transforming growth factor, beta receptor III, transforming growth factor, beta receptor III (betaglycan, 300kDa) |
| Gene Symbols | TGFBR3 |
| Host Species | Sheep |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
