Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGIF1 Antibody (OTI1B12), FITC, Novus Biologicals™
SDP

Catalog No. NBP274496F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274496F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274496F Supplier Novus Biologicals Supplier No. NBP274496F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TGIF1 Monoclonal specifically detects TGIF1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.

Specifications

Antigen TGIF1
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI1B12
Conjugate FITC
Dilution Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias 5′-TG-3′ interacting factor, homeobox protein TGIF, homeobox protein TGIF1, HPE4, MGC39747, TALE homeobox TG-interacting factor, TGFB-induced factor (TALE family homeobox), TGFB-induced factor homeobox 1, TGIFMGC5066,5′-TG-3′-interacting factor 1, transforming growth factor-beta-induced factor
Gene Symbols TGIF1
Host Species Mouse
Immunogen Full length recombinant protein of human TGIF1 (NP_775300) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7050.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.