Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ TGN 020

Click to view available options
Quantity:
10 mg
50 mg
Description
TGN 020 is an aquaporin 4 (AQP4) channel blocker (IC50 = 3.1 μM). Increases regional cerebral blood flow and reduces ischemia-induced brain edema in mice.
Specifications
Specifications
Quantity | 10 mg |
Product Type | TGN 020 |
Purity | >99% |
Molecular Weight (g/mol) | 206.22 |
Inhibitors | TGN 020 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction