Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGN38 Antibody (2F7.1), Novus Biologicals™
SDP

Catalog No. NB300575 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300575 100 μL
NB300575SS 0.02 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB300575 Supplier Novus Biologicals Supplier No. NB300575
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 9 publications

TGN38 Monoclonal specifically detects TGN38 in Human, Mouse, Rat, Chinese Hamster, Monkey, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen TGN38
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Neutralization
Classification Monoclonal
Clone 2F7.1
Conjugate Unconjugated
Dilution Western Blot 1:500, Flow Cytometry 1:20 - 1:50, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 1:1000
Gene Alias TGN46TGN38 homolog, TGN48MGC14722, TGN5148, 51kD isoforms), trans-golgi network protein 2, Trans-Golgi network protein TGN51
Gene Symbols TGOLN2
Host Species Mouse
Immunogen Synthetic peptide corresponding to residues L(1) P S A S K P N N T S S E N N P P(17) C of rat TGN38.
Purification Method Protein A or G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Golgi Apparatus Markers, Trans golgi Network Markers
Primary or Secondary Primary
Gene ID (Entrez) 10618
Test Specificity Detects Trans Golgi Network (TGN) 38 from human and monkey samples and both recombinant and endogenous rat samples.
Target Species Human, Mouse, Rat, Chinese Hamster, Monkey, Rabbit
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.