Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGN38 Antibody (2F7.1), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 9 publications

$244.50 - $548.00

Specifications

Antigen TGN38
Clone 2F7.1
Dilution Western Blot 1:500, Flow Cytometry 1:20 - 1:50, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 1:1000
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Neutralization
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB300575
SDP
View Documents
Novus Biologicals
NB300575
100 μL
Each for $548.00
Only null left
Add to Cart
 
NB300575SS
SDP
View Documents
Novus Biologicals
NB300575SS
0.02 mL
Each for $244.50
Only null left
Add to Cart
 
Description

Description

TGN38 Monoclonal specifically detects TGN38 in Human, Mouse, Rat, Chinese Hamster, Monkey, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

TGN38
Western Blot 1:500, Flow Cytometry 1:20 - 1:50, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 1:1000
Monoclonal
Ascites
RUO
Human, Mouse, Rat, Chinese Hamster, Monkey, Rabbit
10618
Synthetic peptide corresponding to residues L(1) P S A S K P N N T S S E N N P P(17) C of rat TGN38.
Primary
Store at -20C. Avoid freeze-thaw cycles.
2F7.1
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Neutralization
Unconjugated
Mouse
Golgi Apparatus Markers, Trans golgi Network Markers
TGN46TGN38 homolog, TGN48MGC14722, TGN5148, 51kD isoforms), trans-golgi network protein 2, Trans-Golgi network protein TGN51
TGOLN2
IgG1
Protein A or G purified
Detects Trans Golgi Network (TGN) 38 from human and monkey samples and both recombinant and endogenous rat samples.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.