Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGN38 Antibody, Novus Biologicals™
SDP

Catalog No. NBP120263 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP120263 0.1 mL
NB018407 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP120263 Supplier Novus Biologicals Supplier No. NBP120263
Add to Cart
Add to Cart

Sheep Polyclonal Antibody has been used in 3 publications

TGN38 Polyclonal antibody specifically detects TGN38 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.

Specifications

Antigen TGN38
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence
Formulation Lyophilized from whole antisera with No Preservative
Gene Accession No. Q62314
Gene Alias TGN46TGN38 homolog, TGN48MGC14722, TGN5148, 51kD isoforms), trans-golgi network protein 2, Trans-Golgi network protein TGN51
Gene Symbols TGOLN2
Host Species Sheep
Immunogen A synthetic peptide from mouse TGN38 conjugated to blue carrier protein was used as the antigen.
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Golgi Apparatus Markers, Trans golgi Network Markers
Primary or Secondary Primary
Gene ID (Entrez) 10618
Test Specificity TGN38 (or TGN46, TGN48, TGN51 in human).
Reconstitution Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the sample size is provided in reconstituted format.
Target Species Human, Mouse, Rat, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.