Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGN46 Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP149643R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP149643R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP149643R Supplier Novus Biologicals Supplier No. NBP149643R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TGN46 Polyclonal specifically detects TGN46 in Human, Mouse, Bovine, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting, Flow (Intracellular), Knockout Validated.

Specifications

Antigen TGN46
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot, Immunoassay
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting, Flow (Intracellular), Knockout Validated
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. O43493
Gene Alias TGN 46, TGN 48, TGN 51, TGN38, TGN-38, TGN38 homolog, TGN46, TGN-46, TGN48, TGN51, TGOLN 2, TGOLN2, trans golgi network 38, trans golgi network 46, Trans Golgi network integral membrane protein 2, Trans Golgi network integral membrane protein 2 [Precursor], Trans Golgi network protein (46 48 51kD isoforms), Trans golgi network protein 2, Trans Golgi network protein TGN51, Trans-Golgi network integral membrane protein 2, Trans-Golgi network protein 2, Trans-Golgi network protein TGN51, TTGN 2, TTGN2
Gene Symbols TGOLN2
Host Species Rabbit
Immunogen A genomic peptide made to an internal region of the human TGN46 protein (within residues 200-350). [Swiss-Prot: O43493]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Golgi Apparatus Markers, Trans golgi Network Markers
Primary or Secondary Primary
Gene ID (Entrez) 10618
Target Species Human, Mouse, Bovine, Monkey
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.