Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TGN46 Mouse anti-Human, Porcine, Rat, Clone: 1F6D5, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | TGN46 |
|---|---|
| Clone | 1F6D5 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6900
![]()
|
Proteintech Group Inc
664771IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
IASPP is one of the most evolutionarily conserved inhibitors of p53 (TP53; MIM 191170), whereas ASPP1. and ASPP2. are activators of p53.Specifications
| TGN46 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| TGN38, TGN38 homolog, TGN46, TGN48, TGN51, TGOLN2, trans Golgi network protein 2, TTGN2 | |
| TGOLN2 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1F6D5 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Pig, Rat | |
| O43493 | |
| 100519476, 10618, 192152 | |
| TGN46 Fusion Protein ag4470 | |
| Primary | |
| -20°C | |
| TGOLN2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title