Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGR5/GPBAR1 Antibody (409522), FITC, Novus Biologicals™
SDP

Catalog No. NB370383 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB370383 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB370383 Supplier Novus Biologicals Supplier No. FAB4286F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TGR5/GPBAR1 Monoclonal specifically detects TGR5/GPBAR1 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen TGR5/GPBAR1
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 409522
Conjugate FITC
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias BG37hBG37, G protein-coupled bile acid receptor 1, GPCR19, GPR131, G-protein coupled bile acid receptor 1, G-protein coupled bile acid receptor BG37, G-protein coupled receptor GPCR19, M-BARhGPCR19, membrane bile acid receptor, Membrane-type receptor for bile acids, MGC40597, TGR5GPCR
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human TGR5/GPBAR1, Met1-Asn330, Accession # Q8TDU6
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 151306
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.