Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGR5/GPBAR1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP223669 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223669 0.1 mL
NBP223669SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP223669 Supplier Novus Biologicals Supplier No. NBP223669
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

TGR5/GPBAR1 Polyclonal specifically detects TGR5/GPBAR1 in Human, Mouse, Rat samples. It is validated for IF/IHC, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), KnockDown, Western Blot.

Specifications

Antigen TGR5/GPBAR1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), KnockDown, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1 - 2 ug/ml, Immunohistochemistry 1:200-1:500, Immunocytochemistry/Immunofluorescence 2 ug/ml, Immunohistochemistry-Paraffin 1:200-1:500, KnockDown Validated
Formulation Whole antisera with 0.05% Sodium Azide
Gene Accession No. Q8TDU6
Gene Alias BG37hBG37, G protein-coupled bile acid receptor 1, GPCR19, GPR131, G-protein coupled bile acid receptor 1, G-protein coupled bile acid receptor BG37, G-protein coupled receptor GPCR19, M-BARhGPCR19, membrane bile acid receptor, Membrane-type receptor for bile acids, MGC40597, TGR5GPCR
Gene Symbols GPBAR1
Host Species Rabbit
Immunogen A portion of amino acid 250-300 of human BG37/TGR5 was used as the immunogen.
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 151306
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, 94% chimpanzee, orangutan and rhesus monkey, 84% in horse and 68% homologous in mouse.
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.