Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TGR5/GPBAR1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$281.00 - $608.50
Specifications
Antigen | TGR5/GPBAR1 |
---|---|
Dilution | Western Blot 1 - 2 ug/ml, Immunohistochemistry 1:200-1:500, Immunocytochemistry/Immunofluorescence 2 ug/ml, Immunohistochemistry-Paraffin 1:200-1:500, KnockDown Validated |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP223669SS
![]() |
Novus Biologicals
NBP223669SS |
0.025 mL |
Each for $281.00
|
|
|||||
NBP223669
![]() |
Novus Biologicals
NBP223669 |
0.1 mL |
Each for $608.50
|
|
|||||
Description
TGR5/GPBAR1 Polyclonal specifically detects TGR5/GPBAR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
TGR5/GPBAR1 | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
Unconjugated | |
Rabbit | |
GPCR | |
Whole antisera with 0.05% Sodium Azide | |
BG37hBG37, G protein-coupled bile acid receptor 1, GPCR19, GPR131, G-protein coupled bile acid receptor 1, G-protein coupled bile acid receptor BG37, G-protein coupled receptor GPCR19, M-BARhGPCR19, membrane bile acid receptor, Membrane-type receptor for bile acids, MGC40597, TGR5GPCR | |
GPBAR1 | |
IgG | |
Unpurified | |
The amino acid sequence used as immunogen is 100% homologous in human, 94% chimpanzee, orangutan and rhesus monkey, 84% in horse and 68% homologous in mouse. |
Western Blot 1 - 2 ug/ml, Immunohistochemistry 1:200-1:500, Immunocytochemistry/Immunofluorescence 2 ug/ml, Immunohistochemistry-Paraffin 1:200-1:500, KnockDown Validated | |
Polyclonal | |
Antisera | |
RUO | |
Human, Mouse, Rat | |
Q8TDU6 | |
151306 | |
A portion of amino acid 250-300 of human BG37/TGR5 was used as the immunogen. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title